Would you like to log in again?
Human Recombinant Sentrin-specific Protease 8 (from E. coli)
Supplier: ProSci
91-152
75789-020EA 389.22 USD InStock
75789-020
Human Recombinant Sentrin-specific Protease 8 (from E. coli)
Enzymes
Sentrin-Specific Protease 8 (SENP8) mediates the reversible covalent modification of proteins by NEDD8
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
SENP8 catalyzes the full-length NEDD8 to generate its mature form and deconjugation of NEDD8 from targeted proteins such as CUL2 , CUL4A in vivo, or p53. but it does not show activity against ubiquitin or SUMO proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty