Would you like to log in again?
Human Recombinant Aldolase A (from E. coli)
Supplier: ProSci
91-182
75789-080EA 700.27 USD InStock
75789-080
Human Recombinant Aldolase A (from E. coli)
Enzymes
Fructose Bisphosphate Aldolase A (ALDOA) belongs to the class I fructose-bisphosphate aldolase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
ALDOA is a glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. In vertebrates, three forms of this ubiquitous glycolytic enzyme are found, Aldolase A in muscle, Aldolase B in liver and aldolase C in brain. Aldolase A Interacts with SNX9 and WAS. Aldolase A deficiency has been associated with myopathy and hemolytic anemia. In addition, Aldolase A plays an important role in glycolysis and gluconeogenesis; it may also act as a scaffolding protein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty