Would you like to log in again?
Human Recombinant Fumarase (from E. coli)
Supplier: ProSci
91-204
75789-124EA 946.47 USD InStock
75789-124
Human Recombinant Fumarase (from E. coli)
Enzymes
Fumarase is an enzyme that catalyze the reversible hydration/dehydration of fumarate to S-malate and is involved in the tricarboxylic acid or Krebs cycle
This recombinant protein can be used for biological assays. For research use only.
Fumarase exists in both form, cytosolic formand N-terminal extend mitochondrial form. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension is the same form as in the cytoplasm. Fumarase is thought to act as a tumor suppressor, which deficiency can lead to progressive encephalopathy, cerebral atrophy and development delay.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty