Would you like to log in again?
Human Recombinant GGPPSase (from E. coli)
Supplier: ProSci
91-209
75789-134EA 946.47 USD InStock
75789-134
Human Recombinant GGPPSase (from E. coli)
Enzymes
Geranylgeranyl pyrophosphate synthase (GGPS1) is a member of the FPP/GGPP synthase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
GGPS1 is highly expressed in testis, heart and skeletal muscle. GGPS1 is localized in the cytoplasm and has geranylgeranyl diphosphate (GGPP) synthase activity. It catalyzes the trans-addition of the three molecules of IPP onto DMAPP to form geranylgeranyl pyrophosphate, an important precursor of carotenoids and geranylated proteins. Other transcriptional splice variants have been found.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty