Would you like to log in again?
Human Recombinant PMKASE (from E. coli)
Supplier: ProSci
91-228
75789-172EA 946.47 USD InStock
75789-172
Human Recombinant PMKASE (from E. coli)
Enzymes
Phosphomevalonate kinase (PMVK) is a cytosolic enzyme
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
PMVK can be highly expressed in the heart,skeletal muscle, liver, pancreas, and kidney; it is expressed at lower levels in the brain, lung, and placenta. Induced by sterol, PMVK takes part in isopentenyl diphosphate biosynthesis through the mevalonate pathway. PMVK catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate in the fifth reaction of the cholesterol biosynthetic pathway.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty