Would you like to log in again?
Human Recombinant Phosphomannomutase 2 (from E. coli)
Supplier: ProSci
91-230
75789-176EA 946.47 USD InStock
75789-176
Human Recombinant Phosphomannomutase 2 (from E. coli)
Enzymes
Phosphomannomutase 2 (PMM2) is an enzyme that is a member of the highly variable methyltransferase superfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
PMM2 is a cytoplasmic protein and catalyzes the isomerization of mannose 6-phosphate to mannose 1-phosphate.In addition, PMM2 involved in the synthesis of the GDP-mannose and dolichol-phosphate-mannose that required for a number of critical mannosyl transfer reactions. Defects in PMM2 can results in congenital disorder of glycosylation type 1A (CDG1A). Congenital disorders of glycosylation are metabolic deficiencies in glycoprotein biosynthesis that usually cause severe mental and psychomotor retardation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty