Would you like to log in again?
Human Recombinant L-Xylulose Reductase (from E. coli)
Supplier: ProSci
91-257
75789-190EA 946.47 USD InStock
75789-190
Human Recombinant L-Xylulose Reductase (from E. coli)
Enzymes
L-Xylulose Reductase is an enzyme that belongs to the Short-Chain Dehydrogenases/Reductases (SDR) family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
L-Xylulose Reductase is responsible for the metabolism of Xylulose, converting it into Xylitol. L-Xylulose Reductase catalyzes the NADPH-dependent reduction of several Pentoses, Tetroses, Trioses, alpha-Dicarbonyl compounds and L-Xylulose. L-Xylulose Reductase participates in the Uronate Cycle of Glucose metabolism. It may play a role in the water absorption and cellular osmoregulation in the proximal renal tubules by producing Xylitol, an osmolyte, thereby preventing osmolytic stress from occurring in the renal tubules.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty