Would you like to log in again?
Human Recombinant Galactose Mutarotase (from E. coli)
Supplier: ProSci
91-258
75789-192EA 946.47 USD InStock
75789-192
Human Recombinant Galactose Mutarotase (from E. coli)
Enzymes
Galactose Mutarotase (GALM) is a cytoplasmic enzyme that belongs to the Aldose Epimerase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
GALM is a Mutarotase that converts alpha-Aldose to the beta -Anomer. GALM is active on D-Glucose, L-Arabinose, D-Xylose, D-Galactose, Maltose and Lactose. GALM may be required for normal Galactose metabolism by maintaining the equilibrium of alpha- and beta - anomers of Galactose.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty