Would you like to log in again?
Human Recombinant beta-Galactosidase (from Cells)
Supplier: ProSci
91-428
75789-532EA 785.03 USD InStock
75789-532
Human Recombinant beta-Galactosidase (from Cells)
Enzymes
beta Galactosidase is a lysosomal beta Galactosidase that hydrolyzes the terminal beta Galactose from Ganglioside and Keratan sulfate
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
In lysosome, the mature beta Galactosidase protein associates with Cathepsin A and Neuraminidase 1 to form the lysosomal multienzyme complex . An alternative splicing at the RNA level of beta Galactosidase results a catalytically inactive beta Galactosidase that plays an important role in vascular development. Defects of beta -galactosidase (GLB1) are the cause of diseases like GM1-gangliosidosis which is a lysosomal storage disease and Morquio Syndrome B that cause patients to have abnormal elastic fibers. More than 100 mutations have been identified for beta Galactosidase, which result in different residual activities of the mutant enzymes and a spectrum of symptoms in the two related diseases.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty