Would you like to log in again?
Human Recombinant Tryptase beta-2 (from Cells)
Supplier: ProSci
91-456
75789-588EA 946.47 USD InStock
75789-588
Human Recombinant Tryptase beta-2 (from Cells)
Enzymes
Tryptases are Trypsin-like Serine Proteases
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
beta -Tryptases are the main isoenzymes in mast cells. beta tryptases form active tetramers with heparin proteoglycan. In the tetramer, the unique arrangement of the active sites facing a narrow central pore, beta -Tryptases are resistant to macromolecule protease inhibitors . When mast cells are activated, beta -Tryptases are released and participate in provoking inflammatory conditions . beta -Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic disorders.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty