Would you like to log in again?
Human Recombinant Protein Disulfide-Isomerase A6 (from Cells)
Supplier: ProSci
91-476
75789-628EA 700.27 USD InStock
75789-628
Human Recombinant Protein Disulfide-Isomerase A6 (from Cells)
Enzymes
Protein Disulfide-Isomerase A6 (PDIA6) is a 48.5kDa protein that belongs to the protein disulfide isomerase family (PDI)
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
PDIA6 is an enzyme in the endoplasmic reticulum in eukaryotes which catalyzes the formation and breakage of disulfide bonds between cysteine residues within proteins as they fold. The PDIA6 expressed in platelets, its functions as a chaperone that inhibits aggregation of misfolded proteins. PDIA6 is part a large chaperone multiprotein complex comprising DNAJB11, HSP90B1, HSPA5, HYOU, PDIA2, PDIA4, PDIA6, PPIB, SDF2L1, UGT1A1. PDIA6 also plays a role in platelet aggregation and activation by agonists such as convulxin, collagen and thrombin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty