Would you like to log in again?
Human Recombinant Alkaline Phosphatase (from Cells)
Supplier: ProSci
91-503
75789-682EA 946.47 USD InStock
75789-682
Human Recombinant Alkaline Phosphatase (from Cells)
Enzymes
Alkaline Phosphatase, Tissue-Nonspecific Isozyme (ALPL) is a cell membrane protein which belongs to the alkaline phosphatase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
There are at least four distinct but related alkaline phosphatases in humans: intestinal AP (IAP), placental AP(PLAP), germ cell AP (GCAP) and their genes are clustered on chromosome 2, tissue-nonspecific isozyme (TNAP) which gene is located on chromosome 1. Alkaline phosphatases (APs) are dimeric enzymes, it catalyze the hydrolysis of phosphomonoesters with release of inorganic phosphate. The native ALPL is a glycosylated homodimer attached to the membrane through a GPI-anchor. This isozyme may play a role in skeletal mineralization. Mutations in ALPL gene have been linked directly to different forms of hypophosphatasia,characterized by poorly mineralized cartilage and bones, and this disorder can vary depending on the specific mutation since this determines age of onset and severity of symptoms.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty