Would you like to log in again?
Human Recombinant Chitinase 3-Like 2 (from Cells)
Supplier: ProSci
91-523
75789-722EA 744.04 USD InStock
75789-722
Human Recombinant Chitinase 3-Like 2 (from Cells)
Enzymes
Chitinase 3-Like Protein 2 (CHI3L2) is a 39 kDa secreted glycoprotein which belongs to the glycosyl hydrolase 18 family and the most closely related to human cartilage glycoprotein 39, which promotes the growth of human synovial cells as well as skin and fetal lung fibroblasts
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Highest expression of CHI3L2 is in chondrocytes, followed by synoviocytes, lung and heart. It is not detected in spleen, pancreas, and liver. CHI3L2 may also be expressed in developing brain and placenta. Increased levels of CHI3L2 have been demonstrated in synovial fluids of patients with rheumatoid or osteoarthritis as well as in some other pathologies and in malignant tumors, particularly in glioblastomas. CHI3L2 may bind glycan structure with high affinity, but not heparin. It has has no chitotriosidase activity, but is likely to bind some type of glycan.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty