Would you like to log in again?
Human Recombinant Proteinase 3 (from Cells)
Supplier: ProSci
91-542
75789-760EA 700.27 USD InStock
75789-760
Human Recombinant Proteinase 3 (from Cells)
Enzymes
Proteinase-3 is a neutral serine proteinase that belongs to the peptidase S1 family and Elastase subfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It contains one peptidase S1 domain and it is expressed mainly in neutrophil granulocytes. The primary function of Proteinase-3 is thought to be degradation of extracellular proteins at sites of inflammation, but excessive or prolonged proteolytic activity may cause harmful effects in the body. It is the epitope of anti-neutrophil cytoplasmic antibodies (ANCAs) of the cANCA (cytoplasmic subtype) class, a type of antibody frequently found in the disease Wegener's granulomatosis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty