Would you like to log in again?
Mouse Recombinant Carboxypeptidase M
Supplier: ProSci
91-622
75789-904EA 389.22 USD InStock
75789-904
Mouse Recombinant Carboxypeptidase M
Enzymes
Carboxypeptidase M (CPM) belongs to the peptidase M14 family, and exists in cell membrane
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
The protein binds 1 zinc ion per subunit, and cleavage of C-terminal arginine or lysine residues from polypeptides. CPM specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty