Would you like to log in again?
Human Recombinant Angiopoietin-like 8 (from Cells)
Supplier: ProSci
91-639
75789-938EA 912.46 USD InStock
75789-938
Human Recombinant Angiopoietin-like 8 (from Cells)
Proteins and Peptides
The protein specifically promotes pancreatic beta cell proliferation and beta cell mass expansion, thereby improving glucose tolerance. It promotes pancreatic beta cell proliferation without insulin resistance. Also it acts as a blood lipid regulator by regulating serum triglyceride levels and possibly by promoting ANGPTL3 cleavage. It interacts with ANGPTL3. It predominantly expressed in liver and also expressed in adipose tissues. The ability of the protein to induce pancreatic beta cell proliferation is promising in diabetes therapy. Betatrophin treatment could supply or replace insulin injections by increasing the number of insulin-producing cells in diabetes.
  • Fusion-Tag: N-Fc tag
This recombinant protein can be used for biological assays. For research use only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty