Would you like to log in again?
Human Recombinant Transaldolase (from Cells)
Supplier: ProSci
91-656
75789-972EA 946.47 USD InStock
75789-972
Human Recombinant Transaldolase (from Cells)
Enzymes
Transaldolase (TALDO1) belongs to the transaldolase family of Type 1 subfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
TALDO1 is expressed selectively in oligodendrocytes of the brain. TALDO1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. TALDO1 deficiency results in telangiectases of the skin, hepatosplenomegaly and enlarged clitoris.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty