Would you like to log in again?
Human Recombinant GMP Reductase 1 (from Cells)
Supplier: ProSci
91-666
75789-992EA 700.27 USD InStock
75789-992
Human Recombinant GMP Reductase 1 (from Cells)
Enzymes
GMP Reductase 1 (GMPR) is a member of the IMPDH/GMPR family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
GMPR exists as a homotetramer and catalyzes the irreversible NADPH-dependent deamination of GMP to IMP. It functions in the conversion of nucleobase, nucleoside and nucleotide derivatives of G to A nucleotides, and in maintaining the intracellular balance of A and G nucleotides. GMP reductase gene expression may be regulated by MITF. At least two different alleles are known.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty