Would you like to log in again?
Human Recombinant Sialate O-Acetylesterase (from Cells)
Supplier: ProSci
91-729
75790-118EA 946.47 USD InStock
75790-118
Human Recombinant Sialate O-Acetylesterase (from Cells)
Enzymes
Sialate O-Acetylesterase (SIAE) belongs to the family of hydrolases, specifically those acting on carboxylic ester bonds
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
SIAE is widely expressed with high expression levels in the testis, prostate, and colon. SIAE catalyzes N-acetyl-O-acetylneuraminate and H2O to N-acetylneuraminate and acetate. SIAE removes O-acetyl ester groups from position 9 of the parent sialic acid, N-acetylneuraminic acid. SIAE down-regulates B lymphocyte antigen receptor signaling (involving CD22), and is required for immunological tolerance. Loss of function mutations in SIAE are much more frequently found in humans with autoimmune diseases especially rheumatoid arthritis and type 1 diabetes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty