Would you like to log in again?
Human Recombinant Tripeptidyl-Peptidase I (from Cells)
Supplier: ProSci
91-745
75790-142EA 946.47 USD InStock
75790-142
Human Recombinant Tripeptidyl-Peptidase I (from Cells)
Enzymes
Tripeptidyl-Peptidase 1 (TPP1) belongs to the peptidase S53 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
TPP1 is detected in all tissues examined with highest levels in heart and placenta and relatively similar levels in other tissues. TPP1 is lysosomal serine protease with tripeptidyl-peptidase I activity. TPP1 may act as a non-specific lysosomal peptidase which generates tripeptides from the breakdown products produced by lysosomal proteinases. TPP1 requires substrates with an unsubstituted N-terminus. TPP1 mutations have also been shown to cause neuronal ceroid lipofuscinosis type 2 (CLN2).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty