Would you like to log in again?
Human Recombinant Lysosomal Pro-X Carboxypeptidase (from Cells)
Supplier: ProSci
91-746
75790-144EA 946.47 USD InStock
75790-144
Human Recombinant Lysosomal Pro-X Carboxypeptidase (from Cells)
Enzymes
Lysosomal Pro-X Carboxypeptidase (PRCP) belongs to the peptidase S28 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
PRCP is detected in many tissues, with highest levels observed in placenta, lung, and liver. It is also present in the heart, brain, pancreas, and kidney. PRCP exists as a homodimer. PRCP cleaves C-terminal amino acids linked to proline in peptides such as angiotensin II, III and des-Arg9-bradykinin. This cleavage occurs at acidic pH, but enzymatic activity is retained with some substrates at neutral pH. PRCP has been shown to be an activator of the cell matrix-associated prekallikrein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty