Would you like to log in again?
Human Recombinant Prostaglandin-D2 Synthase (from Cells)
Supplier: ProSci
91-757
75790-166EA 700.27 USD InStock
75790-166
Human Recombinant Prostaglandin-D2 Synthase (from Cells)
Enzymes
Prostaglandin-H2 D-Isomerase (PTGDS) belongs to the Lipocalin family of calycin superfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
PTGDS is preferentially expressed in the brain. PTGDS catalyzes the conversion of PGH2 to PGD2, a prostaglandin involved in smooth muscle contraction/relaxation and a potent inhibitor of platelet aggregation. PTGDS is involved in a variety of CNS functions, such as sedation, REM sleep and PGE2-induced allodynia, and may have an anti-apoptotic role in oligodendrocytes. PTGDS binds small non-substrate lipophilic molecules and may act as a scavenger for harmful hydrophopic molecules and a secretory retinoid and thyroid hormone transporter. It possibly participates in development and maintenance of the blood-brain, blood-retina, blood-aqueous humor, blood-testis barrier, the central nervous system and male reproductive system.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty