Would you like to log in again?
Human Recombinant Pancreatic Lipase-Related 2 (from Cells)
Supplier: ProSci
91-758
75790-168EA 700.27 USD InStock
75790-168
Human Recombinant Pancreatic Lipase-Related 2 (from Cells)
Enzymes
Pancreatic Lipase-Related Protein 2 (PNLIPRP2) is a secreted protein that belongs to the Lipase family of AB hydrolase superfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
PNLIPRP2 is a lipase with broad substrate specificity that contains one PLAT domain. PNLIPRP2 is expressed mainly in pancreas. PNLIPRP2 can hydrolyze both phospholipids and galactolipids. PNLIPRP2 acts preferentially on monoglycerides, phospholipids and galactolipids. PNLIPRP2 also contributes to milk fat hydrolysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty