Would you like to log in again?
Human Recombinant Alkaline Phosphatase (from Cells)
Supplier: ProSci
91-795
75790-242EA 946.47 USD InStock
75790-242
Human Recombinant Alkaline Phosphatase (from Cells)
Enzymes
ALPP is a membrane protein and exits as a homodimer
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
ALPP is expressed only in normal term placenta, endocervix and fallopian tube and also in ovarian and proximal gastrointestinal tumors. It has been shown to play a role in a number of processes including cell signaling, long-term potentiation, and cell adhesion, however, the best known and most commonly studied role is implicated in Alzheimer's research.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty