Would you like to log in again?
Human Recombinant Protein disulfide-isomerase A3 (from Cells)
Supplier: ProSci
91-869
75790-374EA 946.47 USD InStock
75790-374
Human Recombinant Protein disulfide-isomerase A3 (from Cells)
Enzymes
PDIA3 protein is also known as Protein disulfide-isomerase A3
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is a protein that in humans is encoded by the PDIA3 gene.PDIA3 is an enzyme that belongs to the endoplasmic reticulum and interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. PDIA3 interacts with thiazide-sensitive sodium-chloride cotransporter in the kidney and is induced by glucose deprivation. PDIA3 is part of the major histocompatibility complex (MHC) class I peptide-loading complex (TAP1), which is important for formation of the final antigen conformation and export from the endoplasmic reticulum to the cell surface.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty