Would you like to log in again?
Human Recombinant Creatine Kinase (from Cells)
Supplier: ProSci
91-873
75790-382EA 531.98 USD InStock
75790-382
Human Recombinant Creatine Kinase (from Cells)
Enzymes
Creatine kinase M-type is also known as Creatine kinase M chain,M-CK
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is a protein that in humans is encoded by the CKM gene. It belongs to the ATP:guanido phosphotransferase family,containing 1 phosphagen kinase C-terminal domain and 1 phosphagen kinase N-terminal domain. Creatine kinase M-type can reversibly catalyzes the transfer of phosphate between ATP and various phosphogens. It plays a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty