Would you like to log in again?
Human Recombinant Fructose-1,6-Bisphosphatase 1 (from Cells)
Supplier: ProSci
91-948
75790-532EA 644.75 USD InStock
75790-532
Human Recombinant Fructose-1,6-Bisphosphatase 1 (from Cells)
Enzymes
Fructose-1,6-bisphosphatase 1(FBP1) is a homotetramer protein and belongs to the FBPase class 1 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It involves in carbohydrate biosynthesis; gluconeogenesis pathway. FBP1 is a gluconeogenesis regulatory protein which catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. FBP1 deficiency is associated with hypoglycemia and metabolic acidosis. FBP1 regulates mouse endogenous glucose production. FBP1 coupled with phosphofructokinase (PFK) takes part in the metabolism of pancreatic islet cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty