Would you like to log in again?
Human Recombinant Protein disulfide-isomerase A5 (from Cells)
Supplier: ProSci
91-965
75790-566EA 946.47 USD InStock
75790-566
Human Recombinant Protein disulfide-isomerase A5 (from Cells)
Enzymes
Protein disulfide-isomerase A5 is a 519 amino acids protein that belongs to the protein disulfide isomerase family.It contains 3 thioredoxin domains.It can catalyze the rearrangement of -S-S- bonds in proteins.
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty