Would you like to log in again?
Human Recombinant Glutamine Synthetase (from E. coli)
Supplier: ProSci
91-970
75790-576EA 946.47 USD InStock
75790-576
Human Recombinant Glutamine Synthetase (from E. coli)
Enzymes
Glutamine Synthetase reglutes intracellular concentration of glutamate
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Glutamine Synthetase catalyzes the synthesis of glutamine from glutamate and ammonia. Glutamine is an important source of energy and that takes part in cell prolifetation, inhibition of apoptosis, and cell signaling. Glutamine Synthetase is expressed during early fetal stages, and has a role in maintaining body PH by removing ammonia from circulation. Mutations in the GLUL gene are related to congenital glutamine deficiency.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty