Would you like to log in again?
Human Recombinant IPP Isomerase 2 (from E. coli)
Supplier: ProSci
92-015
75790-624EA 946.47 USD InStock
75790-624
Human Recombinant IPP Isomerase 2 (from E. coli)
Enzymes
Isopentenyl Pyrophosphate Isomerase 2 (IDI2) belongs to the IPP isomerase type 1 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Both isozymes, IDI1 and IDI2 are localized to the peroxisome by a PTS1-dependent pathway. IDI2 is expressed in skeletal muscle, which contains one nudix hydrolase domain. IDI2 binds one magnesium per subunit. IDI2 catalyzes the 1,3-allylic rearrangement of the homoallylic substrate isopentenyl (IPP) to its highly electrophilic allylic isomer, dimethylallyl diphosphate (DMAPP). It is reported that IDI2 is regulated independently from IDI1, by a mechanism that may involve PPAR- alpha.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty