Would you like to log in again?
Human Recombinant Carbonic Anhydrase 10 (from E. coli)
Supplier: ProSci
92-058
75790-710EA 744.04 USD InStock
75790-710
Human Recombinant Carbonic Anhydrase 10 (from E. coli)
Enzymes
Carbonic Anhydrase-Related Protein 10 (CA10) belongs to the Carbonic Anhydrase family of Zinc Metalloenzymes
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
It is an acatalytic member of the alpha-carbonic anhydrase subgroup. CA10 expression is detected in the adult total brain and almost all parts of the central nervous system, but not in the fetal brain. CA10 catalyze the reversible hydration of carbon dioxide in various biological processes, which is fundamental to many processes such as respiration, renal tubular acidification and bone resorption. It is thought to play a role in the central nervous system, especially in brain development.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty