Would you like to log in again?
Human Recombinant dUTP Pyrophosphatase (from E. coli)
Supplier: ProSci
92-088
75790-770EA 744.04 USD InStock
75790-770
Human Recombinant dUTP Pyrophosphatase (from E. coli)
Enzymes
Deoxyuridine 5'-Triphosphate Nucleotidohydrolase Mitochondrial (dUTPase) belongs to the dUTPase family
This recombinant protein can be used for biological assays. For research use only.
dUTPase exits as a homotrimer and is involved in nucleotide metabolism. dUTPase produces dUMP, the immediate precursor of thymidine nucleotides and it decreases the intracellular concentration of dUTP so that uracil cannot be incorporated into DNA. The dUTPase increase in PCR product yield, length and fidelity enables further down-stream applications. These effects make dUTPase useful in PCR fidelity and yield-sensitive applications. dUTPase is specific for dUTP and is critical for the fidelity of DNA replication and repair.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty