Would you like to log in again?
Human Recombinant B Lymphocyte Kinase (from E. coli)
Supplier: ProSci
92-107
75790-808EA 700.27 USD InStock
75790-808
Human Recombinant B Lymphocyte Kinase (from E. coli)
Enzymes
Tyrosine-Protein Kinase Blk (BLK) contains one protein kinase domain, one SH2 domain and one SH3 domain
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
BLK is a non-receptor tyrosine kinase, which is involved in B-lymphocyte development, differentiation and signaling. B-cell receptor (BCR) signaling requires a tight regulation of several protein tyrosine kinases and phosphatases, and associated coreceptors. Signaling through BLK plays an important role in transmitting signals through surface immunoglobulines and supports the pro-B to pre-B transition, as well as the signaling for growth arrest and apoptosis downstream of B-cell receptor. Defects in BLK are a cause of maturity-onset diabetes of the young type 11 (MODY11).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty