Would you like to log in again?
Human Recombinant Cystathionine Gamma-Lyase (from E. coli)
Supplier: ProSci
92-113
75790-820EA 744.04 USD InStock
75790-820
Human Recombinant Cystathionine Gamma-Lyase (from E. coli)
Enzymes
Cystathionine Gamma-Lyase (CTH) belongs to the trans-sulfuration enzymes family
This recombinant protein can be used for biological assays. For research use only.
CTH exists as a homotetramer and interacts with CALM in a calcium-dependent manner. CTH breaks down cystathionine into cysteine, ammonia and 2-oxobutanoate. CTH catalyzes the last step in the trans-sulfuration pathway from methionine to cysteine and has broad substrate specificity. Defects in CTH will lead to cystathioninuria, which is an autosomal recessive phenotype characterized by abnormal accumulation of plasma cystathionine.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty