Would you like to log in again?
Human Recombinant Sulfotransferase 1A3 (from E. coli)
Supplier: ProSci
92-141
75790-876EA 946.47 USD InStock
75790-876
Human Recombinant Sulfotransferase 1A3 (from E. coli)
Enzymes
Sulfotransferase 1A3/1A4 (SULT1A3) is 295 amino acids in length and localizes to the cytoplasm
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
It is a member of the Sulfotransferase 1 family. SULT1A3 can be found in the liver, colon, kidney, lung, brain, spleen, small intestine, placenta, and leukocytes. SULT1A3 exists as a homodimer and it catalyzes the sulfation of phenolic monoamines, such as dopamine, norepinephrine and serotonin, and phenolic and catecholic drugs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty