Would you like to log in again?
Human Recombinant Caspase 14 (from E. coli)
Supplier: ProSci
92-153
75790-900EA 912.19 USD InStock
75790-900
Human Recombinant Caspase 14 (from E. coli)
Enzymes
Caspase 14 (CASP14) is an enzyme that belongs to the peptidase C14A family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
The Caspase 14 protein is complexed of unprocessed caspase-14 and processed 19 kDa (p19) and 10 kDa (p10) subunits. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes, which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. CASP14 has been shown to be processed and activated by Caspase 8 and Caspase 10 in vitro, and by anti-Fas agonist antibody or TNF-related apoptosis inducing ligand in vivo. The expression and processing of this caspase may be involved in keratinocyte terminal differentiation, which is important for the formation of the skin barrier. It is believed to be a non-apoptotic caspase which is involved in epidermal differentiation, keratinocyte differentiation and cornification. CASP14 probably regulates maturation of the epidermis by proteolytically processing filaggrin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty