Would you like to log in again?
Human Recombinant Tryptophan 5-Hydroxylase 2 (from E. coli)
Supplier: ProSci
92-161
75790-916EA 946.47 USD InStock
75790-916
Human Recombinant Tryptophan 5-Hydroxylase 2 (from E. coli)
Enzymes
Tryptophan 5-hydroxylase 2 (TPH2) is an isozyme member of the biopterin-dependent aromatic amino acid hydroxylase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
TPH2 is primarily expressed in the serotonergic neurons of the brain, with the highest expression found in the raphe nucleus of the midbrain. TPH2 catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. Genetic variation in TPH2 may influence susceptibility to major depressive disorder (MDD). Defects in TPH2 are the cause of psychiatric diseases such as bipolar affective disorder and major depression.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty