Would you like to log in again?
Human Recombinant CaM kinase ID (from E. coli)
Supplier: ProSci
92-220
75791-034EA 946.47 USD InStock
75791-034
Human Recombinant CaM kinase ID (from E. coli)
Enzymes
CAMK1D is a serine?threonine kinase that is a member of the calcium?calmodulin-dependent protein kinase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
It contains a protein kinase domain. CAMK1D expressed in polymorphonuclear leukocytes and may be part of the chemokine signal transduction pathway that regulates granulocyte function. It may also be involved in modulation of neuronal apoptosis. It activates CREB-dependent gene transcription, regulates calcium-mediated granulocyte function and respiratory burst and promotes basal dendritic growth of hippocampal neurons.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty