Would you like to log in again?
Human Recombinant ATPase SWSAP1 (from E. coli)
Supplier: ProSci
92-241
75791-076EA 912.46 USD InStock
75791-076
Human Recombinant ATPase SWSAP1 (from E. coli)
Enzymes
SWSAP1 is a nucleus ATPase protein, interacts with ZSWIM7 and forms a functional complex
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
The complexs involved in homologous recombination repair and stabilizes each other. SWS1AP1 also interacts with RAD51, RAD51B, RAD51C, RAD51D and XRCC3. It involves in homologous recombination repair. ATPase is preferentially stimulated by single-stranded DNA and is involved in homologous recombination repair (HRR). SWSAP1 has a DNA-binding activity which is independent of its ATPase activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty