Would you like to log in again?
Human Recombinant Dipeptidyl-peptidase 3 (from E. coli)
Supplier: ProSci
92-263
75791-120EA 946.47 USD InStock
75791-120
Human Recombinant Dipeptidyl-peptidase 3 (from E. coli)
Enzymes
Dipeptidyl peptidase 3(DPP3), is a member of the S9B family in clan SC of the serine proteases
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
DPP3 has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH). It releases an N-terminal dipeptide from a peptide comprising four or more residues, with broad specificity and also acts on dipeptidyl 2-naphthylamides.Increased activity of this protein has a relationship with endometrial and ovarian cancers.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty