Would you like to log in again?
Human Recombinant Methenyl-THF synthetase (from E. coli)
Supplier: ProSci
92-280
75791-154EA 946.47 USD InStock
75791-154
Human Recombinant Methenyl-THF synthetase (from E. coli)
Enzymes
5-formyltetrahydrofolate cyclo-ligase (MTHFS) belongs to the 5-formyltetrahydrofolate cyclo-ligase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is an enzyme that catalyzes the conversion of 5-formyltetrahydrofolate to 5,10-methenyltetrahydrofolate, contributes to tetrahydrofolate metabolism. MTHFS helps regulate carbon flow through the folate-dependent one-carbon metabolic network that supplies carbon for the biosynthesis of purines, thymidine and amino acids. An increased activity of the encoded protein can result in an increased folate turnover rate and folate depletion.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty