Would you like to log in again?
E. coli Recombinant beta-Glucuronidase (from E. coli)
Supplier: ProSci
92-300
75791-186EA 700.27 USD InStock
75791-186
E. coli Recombinant beta-Glucuronidase (from E. coli)
Enzymes
Beta-D-glucuronidase from E
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
coli is a  highly specific enzyme in hydrolyzing glucuronides. Beta-D-glucuronidase (GLUase) activity can be measured as the rate of production of fluorescent methylumbelliferone (MU), resulting from the hydrolysis of the substrate 4-methylumbelliferyl- beta -d-glucuronide (MUGLU), which is an effective and rapid method for detection and verification of E. coli in food, water, and environmental samples. High purity recombinant beta-Glucuronidase is used in research, biochemical enzyme assays and in vitro diagnostic analysis, detecting a wide range of drugs such as opioids, benzodiazepines, steroids, cannabinoids, and others.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty