Would you like to log in again?
Human Recombinant Aminoacylase-3 (from E. coli)
Supplier: ProSci
92-310
75791-206EA 700.27 USD InStock
75791-206
Human Recombinant Aminoacylase-3 (from E. coli)
Enzymes
Aspartoacylase 3, also known as ACY3, N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming), Acylase III, Aminoacylase-3, Aspartoacylase-2, Aspartoacylase-2, HCV core-binding protein 1 and ASPA2, is a member of the Aspartoacylase subfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
ACY3 plays an important role in deacetylating mercapturic acids in kidney proximal tubules and acts on N-acetyl-aromatic amino acids.ACY3 is located in the cytoplasm of S2 and S3 proximal tubules and the apical domain of S1 proximal tubules. ACY3 protein is also expressed at low levels in stomach, testis, heart, brain, lung and liver, and may function as an HCV (Hepatitis C virus) core binding protein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty