Would you like to log in again?
Mouse Recombinant Transglutaminase 2
Supplier: ProSci
92-325
75791-236EA 946.47 USD InStock
75791-236
Mouse Recombinant Transglutaminase 2
Enzymes
Protein-glutamine gamma-glutamyltransferase 2 (TGM2) is a 78-kDa, calcium dependent enzyme,It belongs to the transglutaminase superfamily and transglutaminase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
The protein encoded by this TGM2 gene acts as a monomer, is induced by retinoic acid, and appears to be involved in apoptosis. TGM2 is the autoantigen implicated in celiac disease. Two transcript variants encoding different isoforms have been found for this gene.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty