Would you like to log in again?
Human Recombinant Ribonuclease T2 (from Cells)
Supplier: ProSci
92-334
75791-254EA 946.19 USD InStock
75791-254
Human Recombinant Ribonuclease T2 (from Cells)
Enzymes
RNASET2, also known as RNASE6PL, is short for bonuclease T2
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is a 256 aa. protein which belongs to the RNase T2 family. RNASET2 is a secreted protein, and is higher expressed in the temporal lobe and fetal brain. This protein can be inhibited by Zn2+ and Cu2+. It has ribonuclease activity, with higher activity at acidic pH and is probably involved in lysosomal degradation of ribosomal RNA. It also plays a role in cellular RNA catabolism.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty