Would you like to log in again?
Human Recombinant alpha-Amylase 2B (from Cells)
Supplier: ProSci
92-341
75791-268EA 629.48 USD InStock
75791-268
Human Recombinant alpha-Amylase 2B (from Cells)
Enzymes
Alpha-amylase 2B is an enzyme that in humans is encoded by the AMY2B gene
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Alpha-amylase 2B is a secreted protein and belongs to the glycosyl hydrolase 13 family. It hydrolyzes 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. This gene encodes an amylase isoenzyme produced by the pancreas. Diseases associated with AMY2B include pancreatitis, and thyroiditis, and among its related super-pathways are Metabolic pathways and MPS IIIC - Sanfilippo syndrome C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty