Would you like to log in again?
Human Recombinant Polypeptide GalNac Transferase 3 (from Cells)
Supplier: ProSci
92-355
75791-296EA 946.47 USD InStock
75791-296
Human Recombinant Polypeptide GalNac Transferase 3 (from Cells)
Enzymes
Polypeptide N-acetylgalactosaminyltransferase 3(GALNT3) belongs to the glycosyltransferase 2 family and galNAc-T subfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It expressed in organs that contain secretory epithelial glands and it highly expressed in pancreas, skin, kidney and testis. There are two conserved domains in the glycosyltransferase region: the N-terminal domain (domain A, also called GT1 motif), which is probably involved in manganese coordination and substrate binding and the C-terminal domain (domain B, also called Gal/GalNAc-T motif), which is probably involved in catalytic reaction and UDP-Gal binding .This protein plays a major role in regulating phosphate levels within the body (phosphate homeostasis). Among its many functions, phosphate plays a critical role in the formation and growth of bones in childhood and helps maintain bone strength in adults.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty