Would you like to log in again?
Human Recombinant Phosphoglycerate kinase 1 (from Cells)
Supplier: ProSci
92-357
75791-300EA 946.47 USD InStock
75791-300
Human Recombinant Phosphoglycerate kinase 1 (from Cells)
Enzymes
PGK1 is called phosphoglycerate kinase that involved in a critical energy-producing process known as glycolysis
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Phosphoglycerate kinase helps carry out a chemical reaction that converts a molecule called 1,3-diphosphoglycerate, which is produced during the breakdown of glucose, to another molecule called 3-phosphoglycerate during glycolysis. PGK1 The encoded protein may also act as a cofactor for polymerase alpha.. The protein has been identified as a moonlighting protein based on its ability to perform mechanistically distinct functions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty