Would you like to log in again?
Human Recombinant Thymidine kinase 1 (from Cells)
Supplier: ProSci
92-369
75791-324EA 946.47 USD InStock
75791-324
Human Recombinant Thymidine kinase 1 (from Cells)
Enzymes
Thymidine kinase 1(TK1) belongs to the thymidine kinase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is located in the cytoplasm, and phosphorylated on Ser-13 in mitosis during post-translational modification. Two forms of this protein have been identified in animal cells, one in cytosol TK1 and one in mitochondria TK2. Thymidine kinases have a key function in the synthesis of DNA and thereby in cell division, as they are part of the unique reaction chain to introduce deoxythymidine into the DNA. Activity of the cytosolic enzyme is high in proliferating cells and peaks during the S-phase of the cell cycle, while it is very low in resting cells. TK1 acts as a homotetramer, and can transform thymidime to thymidine 5'-phosphate with the help of ATP
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty