Would you like to log in again?
Human Recombinant Thymidylate kinase (from Cells)
Supplier: ProSci
92-370
75791-326EA 946.47 USD InStock
75791-326
Human Recombinant Thymidylate kinase (from Cells)
Enzymes
Thymidylate kinase (DTYMK) is a ubiquitous enzyme of about 25 kD which belongs to thymidylate kinase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
DTYMK is important in the dTTP synthesis pathway for DNA synthesis. It participated in the pyrimidine metabolism pathway and dTTP biosynthesis pathway. DTYMK catalyzes the conversion of dTMP to dTDP and catalyzes the phosphorylation of thymidine 5'-monophosphate (dTMP) to form thymidine 5'-diphosphate (dTDP) in the presence of ATP and magnesium. Structural and functional analyses suggest that the cDNA codes for authentic human dTMP kinase. The mRNA levels and enzyme activities corresponded to cell cycle progression and cell growth stages.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty